ENA238713026 Enamine

5-(2,4-dichlorophenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD00501904
  • Cas Number : 168893-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51687

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51687
Availability details for
Quantity
  • Availability
  • Add to Cart