ENA238713038 Enamine

1-(3,3-dimethylbutyl)piperazine

  • MDL number MFCD11645196
  • Cas Number : 57184-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...122
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51703

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51703
Availability details for
Quantity
  • Availability
  • Add to Cart