ENA238713063 Enamine

4-[2-(4-methylpiperazin-1-yl)ethyl]aniline

  • MDL number MFCD09884759
  • Cas Number : 262368-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51738
Availability details for
Quantity
  • Availability
  • Add to Cart