ENA238713064 Enamine

2-amino-2-(3-bromophenyl)acetonitrile

  • MDL number MFCD09734172
  • Cas Number : 120667-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51739
Availability details for
Quantity
  • Availability
  • Add to Cart