ENA238713065 Enamine

2-(trifluoromethyl)-[1,1'-biphenyl]-4-amine

  • MDL number MFCD09042387
  • Cas Number : 641570-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51740
Availability details for
Quantity
  • Availability
  • Add to Cart