ENA238713072 Enamine

4-(2,6-dimethylmorpholine-4-carbonyl)-1-methyl-1H-pyrazol-5-amine

  • MDL number MFCD12913012
  • Cas Number : 1181522-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51751
Availability details for
Quantity
  • Availability
  • Add to Cart