ENA238713079 Enamine

ethyl 5-methyl-2-(morpholin-4-yl)-4-oxo-3H,4H-thieno[2,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD07759637
  • Cas Number : 893801-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51775
Availability details for
Quantity
  • Availability
  • Add to Cart