ENA238713083 Enamine

1-(5-methyl-1-phenyl-1H-1,2,3-triazol-4-yl)ethan-1-one

  • MDL number MFCD10003293
  • Cas Number : 51118-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51789
Availability details for
Quantity
  • Availability
  • Add to Cart