ENA238713090 Enamine

5-methyl-1-(4-methylphenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD09035308
  • Cas Number : 210159-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51796
Availability details for
Quantity
  • Availability
  • Add to Cart