ENA238713092 Enamine

6-chloro-1H-1,3-benzodiazole-2-sulfonic acid

  • MDL number MFCD00965565
  • Cas Number : 40828-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 329...331
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51798

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51798
Availability details for
Quantity
  • Availability
  • Add to Cart