ENA238713106 Enamine

1-(4-hydroxypiperidin-1-yl)-2-(methylamino)ethan-1-one hydrochloride

  • MDL number MFCD12913018
  • Cas Number : 1181868-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51818
Availability details for
Quantity
  • Availability
  • Add to Cart