ENA238713126 Enamine

[(2-methoxyphenyl)methyl]urea

  • MDL number MFCD01314650
  • Cas Number : 99362-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51847

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51847
Availability details for
Quantity
  • Availability
  • Add to Cart