ENA238713134 Enamine

4-(1,2,3-thiadiazol-4-yl)aniline

  • MDL number MFCD00052104
  • Cas Number : 121180-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51856
Availability details for
Quantity
  • Availability
  • Add to Cart