ENA238713136 Enamine

N-(2-aminoethyl)-3-methylbenzamide

  • MDL number MFCD09730911
  • Cas Number : 953900-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51861
Availability details for
Quantity
  • Availability
  • Add to Cart