ENA238713137 Enamine

3-(chloromethyl)-N,N-dimethylbenzamide

  • MDL number MFCD11208394
  • Cas Number : 442910-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51862
Availability details for
Quantity
  • Availability
  • Add to Cart