ENA238713138 Enamine

4-acetyl-N-[3-(dimethylamino)propyl]benzene-1-sulfonamide

  • MDL number MFCD12617771
  • Cas Number : 1003494-10-7
  • Molecular Weight : 284.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-51863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51863
Availability details for
Quantity
  • Availability
  • Add to Cart