ENA238713139 Enamine

ethyl 3-(chlorosulfonyl)propanoate

  • MDL number MFCD07186267
  • Cas Number : 103472-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51864

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51864
Availability details for
Quantity
  • Availability
  • Add to Cart