ENA238713157 Enamine

1-phenylpiperidin-4-one

  • MDL number MFCD07367770
  • Cas Number : 19125-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51895
Availability details for
Quantity
  • Availability
  • Add to Cart