ENA238713160 Enamine

2-(1-chloroethyl)-1,3,5-trimethylbenzene

  • MDL number MFCD11186418
  • Cas Number : 51270-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-51898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51898
Availability details for
Quantity
  • Availability
  • Add to Cart