ENA238713164 Enamine

bis(3-methylbutyl)sulfamoyl chloride

  • MDL number MFCD08442618
  • Cas Number : 926230-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51902

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51902
Availability details for
Quantity
  • Availability
  • Add to Cart