ENA238713180 Enamine

5-bromo-2-phenylquinoline-4-carboxylic acid

  • MDL number MFCD08445215
  • Cas Number : 924251-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51931
Availability details for
Quantity
  • Availability
  • Add to Cart