ENA238713184 Enamine

3-(4-aminophenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11167814
  • Cas Number : 22036-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-148
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51935
Availability details for
Quantity
  • Availability
  • Add to Cart