ENA238713200 Enamine

3-tert-butyl-1H-pyrazol-5-amine

  • MDL number MFCD00067987
  • Cas Number : 82560-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78...80
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51959

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-51959
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart