ENA238713203 Enamine

2-bromo-4-methanesulfonylaniline

  • MDL number MFCD09800837
  • Cas Number : 57946-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51962

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51962
Availability details for
Quantity
  • Availability
  • Add to Cart