ENA238713224 Enamine

(1R)-1-(3-fluorophenyl)ethan-1-ol

  • MDL number MFCD03092995
  • Cas Number : 126534-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-51993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-51993
Availability details for
Quantity
  • Availability
  • Add to Cart