ENA238713236 Enamine

4-(4-methylpiperidine-1-carbonyl)aniline

  • MDL number MFCD04035367
  • Cas Number : 79868-20-5
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-52011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52011
Availability details for
Quantity
  • Availability
  • Add to Cart