ENA238713238 Enamine

2-methanesulfonyl-5-methylaniline

  • MDL number MFCD11645433
  • Cas Number : 876494-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52013
Availability details for
Quantity
  • Availability
  • Add to Cart