ENA238713242 Enamine

(1-aminopropan-2-yl)dimethylamine

  • MDL number MFCD00014818
  • Cas Number : 19764-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52019

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-52019
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart