ENA238713256 Enamine

2-(1H-pyrazol-1-yl)ethanethioamide

  • MDL number MFCD09933278
  • Cas Number : 1087353-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-50195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-50195
Availability details for
Quantity
  • Availability
  • Add to Cart