ENA238713258 Enamine

5-(thiophen-3-yl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD11137508
  • Cas Number : 70057-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211...213
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52038

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52038
Availability details for
Quantity
  • Availability
  • Add to Cart