ENA238713264 Enamine

2-(pyrimidin-2-ylsulfanyl)propanoic acid

  • MDL number MFCD08753438
  • Cas Number : 919617-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52044

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52044
Availability details for
Quantity
  • Availability
  • Add to Cart