ENA238713266 Enamine

4-methyl-1-(piperazin-1-yl)pentan-1-one

  • MDL number MFCD11169684
  • Cas Number : 68536-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52046

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52046
Availability details for
Quantity
  • Availability
  • Add to Cart