ENA238713402 Enamine

2-(chloromethyl)-5-ethoxy-1,3,4-thiadiazole

  • MDL number MFCD12913054
  • Cas Number : 4153-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52054
Availability details for
Quantity
  • Availability
  • Add to Cart