ENA238713406 Enamine

ethyl 5-amino-2,3-dihydro-1H-indole-1-carboxylate hydrochloride

  • MDL number MFCD12913058
  • Cas Number : 1193390-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52079
Availability details for
Quantity
  • Availability
  • Add to Cart