ENA238713408 Enamine

2-{1H-pyrrolo[2,3-b]pyridine-3-sulfonamido}acetic acid

  • MDL number MFCD12913062
  • Cas Number : 1193387-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52087
Availability details for
Quantity
  • Availability
  • Add to Cart