ENA238713416 Enamine

5-{[3-(trifluoromethyl)phenyl]methyl}-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD12913072
  • Cas Number : 1193389-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52190
Availability details for
Quantity
  • Availability
  • Add to Cart