ENA238713420 Enamine

4-[(2-phenoxyethyl)amino]butanoic acid hydrochloride

  • MDL number MFCD12913076
  • Cas Number : 1193390-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52201
Availability details for
Quantity
  • Availability
  • Add to Cart