ENA238713423 Enamine

{[5-(furan-2-yl)-1,2-oxazol-3-yl]methyl}(methyl)amine hydrochloride

  • MDL number MFCD12913079
  • Cas Number : 857283-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52209
Availability details for
Quantity
  • Availability
  • Add to Cart