ENA238713426 Enamine

N-[1-(N'-hydroxycarbamimidoyl)cyclopentyl]acetamide

  • Cas Number : 1193390-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-52250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52250
Availability details for
Quantity
  • Availability
  • Add to Cart