ENA238713428 Enamine

N-[1-(N'-hydroxycarbamimidoyl)cycloheptyl]acetamide

  • MDL number MFCD12913086
  • Cas Number : 1193390-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52252
Availability details for
Quantity
  • Availability
  • Add to Cart