ENA238713440 Enamine

N-[4-amino-3-(trifluoromethyl)phenyl]-5-methyl-1,2-oxazole-4-carboxamide hydrochloride

  • MDL number MFCD12913101
  • Cas Number : 1193388-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52365
Availability details for
Quantity
  • Availability
  • Add to Cart