ENA238713443 Enamine

N-(propan-2-yl)-4-propylcyclohexan-1-amine hydrochloride

  • MDL number MFCD12913111
  • Cas Number : 1193389-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52434
Availability details for
Quantity
  • Availability
  • Add to Cart