ENA238713445 Enamine

4-{[(4-chlorophenyl)methyl]amino}butanoic acid hydrochloride

  • MDL number MFCD12913113
  • Cas Number : 1193389-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52451
Availability details for
Quantity
  • Availability
  • Add to Cart