ENA238713446 Enamine

2,3-dihydro-1H-indole-5-sulfonamide hydrochloride

  • MDL number MFCD12913114
  • Cas Number : 1193388-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-52454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52454
Availability details for
Quantity
  • Availability
  • Add to Cart