ENA238713449 Enamine

3-(piperidin-4-yl)propanoic acid hydrochloride

  • MDL number MFCD06202371
  • Cas Number : 51052-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248...250
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52486

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52486
Availability details for
Quantity
  • Availability
  • Add to Cart