ENA238713450 Enamine

2-{[(3-fluorophenyl)methyl]amino}acetic acid hydrochloride

  • MDL number MFCD12913119
  • Cas Number : 1193387-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52527

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52527
Availability details for
Quantity
  • Availability
  • Add to Cart