ENA238713452 Enamine

5-(2-methyl-1,3-thiazol-4-yl)thiophene-3-carboxylic acid

  • MDL number MFCD12913122
  • Cas Number : 1193389-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52533
Availability details for
Quantity
  • Availability
  • Add to Cart