ENA238713453 Enamine

3-methyl-2-(piperidin-1-yl)butanoic acid hydrochloride

  • MDL number MFCD12913123
  • Cas Number : 1214690-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52544
Availability details for
Quantity
  • Availability
  • Add to Cart