ENA238713467 Enamine

5-fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD12913144
  • Cas Number : 223754-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268...270
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52653

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52653
Availability details for
Quantity
  • Availability
  • Add to Cart