ENA238713533 Enamine

1-(1-bromoethyl)-3-chlorobenzene

  • MDL number MFCD11103128
  • Cas Number : 65130-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-52230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52230
Availability details for
Quantity
  • Availability
  • Add to Cart